Tisk…

5-Bromobenzo[d]isoxazole

Dodavatel: Apollo Scientific

OR907213-1G OR907213-5G
APOSOR907213-1GEA 3060 CZK
APOSOR907213-1G APOSOR907213-5G
5-Bromobenzo[d]isoxazole
5-Bromobenzo[d]isoxazole
Vzorec: C₇H₄BrNO
MW: 198,0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659644
CAS číslo: 837392-65-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR