Tisk…

2-(2-Methoxypropan-2-yl)pyrrolidine 97%

Dodavatel: Apollo Scientific

OR907209-1G OR907209-250MG
APOSOR907209-1GEA 13660 CZK
APOSOR907209-1G APOSOR907209-250MG
2-(2-Methoxypropan-2-yl)pyrrolidine 97%
2-(2-Methoxypropan-2-yl)pyrrolidine
Vzorec: C₈H₁₇NO
Storage Temperature: Ambient
CAS číslo: 160142-25-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR