Tisk…

3-Hydroxy-2-(2,2,2-trimethylacetamido)pyridine 95%

Dodavatel: Apollo Scientific

OR907125-1G OR907125-250MG
APOSOR907125-1GEA 18420 CZK
APOSOR907125-1G APOSOR907125-250MG
3-Hydroxy-2-(2,2,2-trimethylacetamido)pyridine 95%
3-Hydroxy-2-(2,2,2-trimethylacetamido)pyridine
Vzorec: C₁₀H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07781130
CAS číslo: 177744-83-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR