(±)-2-Bromovaleric acid 98%
Dodavatel: Apollo Scientific
Synonyms:
(±)-2-Bromopentanoic acid, 2-Bromovalericacid
OR907076-500G
APOSOR907076-500GEA
4400
CZK
APOSOR907076-500G
(±)-2-Bromovaleric acid 98%
(±)-2-Bromovaleric acid
Vzorec:
H₃C(CH₂)₂BrCHCOOH MW: 181,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004217 CAS číslo: 584-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...