2-Chloro-6-hydroxybenzoic acid 95%
Dodavatel: Apollo Scientific
Synonyms:
6-Chloro-2-hydroxybenzoic acid
OR907056-5G
APOSOR907056-5GEA
3640
CZK
APOSOR907056-5G
2-Chloro-6-hydroxybenzoic acid 95%
2-Chloro-6-hydroxybenzoic acid
Vzorec:
C₇H₅ClO₃ MW: 172,5700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10687031 CAS číslo: 56961-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...