2-(4-Dimethylaminophenyl)ethylamine 95%
Dodavatel: Apollo Scientific
OR907037-1G
OR907037-250MG
APOSOR907037-1GEA
20470
CZK
APOSOR907037-1G
APOSOR907037-250MG
2-(4-Dimethylaminophenyl)ethylamine 95%
2-(4-Dimethylaminophenyl)ethylamine
Vzorec:
C₁₀H₁₆N₂ Storage Temperature: Ambient |
CAS číslo:
52632-05-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...