4-Chloro-5-methylpyrimidine 98%
Dodavatel: Apollo Scientific
OR907028-5G
APOSOR907028-5GEA
6020
CZK
APOSOR907028-5G
4-Chloro-5-methylpyrimidine 98%
4-Chloro-5-methylpyrimidine
Vzorec:
C₅H₅ClN₂ MW: 128,5596 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08236809 CAS číslo: 51957-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...