Tisk…

3,4-Dihydropyrrolo[1,2-a]pyrazin-1(2H)-one 98%

Dodavatel: Apollo Scientific

OR906935-100MG OR906935-1G OR906935-250MG
APOSOR906935-100MGEA 870 CZK
APOSOR906935-100MG APOSOR906935-1G APOSOR906935-250MG
3,4-Dihydropyrrolo[1,2-a]pyrazin-1(2H)-one 98%
3,4-Dihydropyrrolo[1,2-a]pyrazin-1(2H)-one
Vzorec: C₇H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD09832035
CAS číslo: 54906-42-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR