Tisk…

2-(Methoxymethyl)piperidine 95%

Dodavatel: Apollo Scientific

OR906842-100MG OR906842-1G OR906842-250MG
APOSOR906842-100MGEA 5390 CZK
APOSOR906842-100MG APOSOR906842-1G APOSOR906842-250MG
2-(Methoxymethyl)piperidine 95%
2-(Methoxymethyl)piperidine
Vzorec: C₇H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD02663567
CAS číslo: 104678-13-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR