2-Chloro-5-methoxy-4-pyridineboronic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
(2-Chloro-5-methoxypyridin-4-yl)boronic acid, 2-Chloro-5-methoxypyridine-4-boronic acid
OR906834-5G
APOSOR906834-5GEA
7180
CZK
APOSOR906834-5G
2-Chloro-5-methoxy-4-pyridineboronic acid 98%
2-Chloro-5-methoxy-4-pyridineboronic acid
Vzorec:
C₆H₇BClNO₃ MW: 187,39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09972106 CAS číslo: 1072945-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...