4-Morpholinocyclohexanone 98%
Dodavatel: Apollo Scientific
OR906652-1G
OR906652-250MG
APOSOR906652-1GEA
4530
CZK
APOSOR906652-1G
APOSOR906652-250MG
4-Morpholinocyclohexanone 98%
4-Morpholinocyclohexanone
Vzorec:
C₁₀H₁₇NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD17013167 CAS číslo: 139025-93-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...