2-Amino-3-methylbenzonitrile 97%
Dodavatel: Apollo Scientific
OR906433-25G
OR906433-5G
APOSOR906433-25GEA
12370
CZK
APOSOR906433-25G
APOSOR906433-5G
2-Amino-3-methylbenzonitrile 97%
2-Amino-3-methylbenzonitrile
Vzorec:
C₈H₈N₂ MW: 132,1625 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09880087 CAS číslo: 69797-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...