Tisk…

5-Chloro-1H-1,2,4-triazole-3-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR906432-1G OR906432-250MG
APOSOR906432-1GEA 22950 CZK
APOSOR906432-1G APOSOR906432-250MG
5-Chloro-1H-1,2,4-triazole-3-carboxylic acid 95%
5-Chloro-1H-1,2,4-triazole-3-carboxylic acid
Vzorec: C₃H₂ClN₃O₂
Storage Temperature: Ambient
CAS číslo: 21733-03-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR