Tisk…

Ethyl 2-(pyridin-3-yloxy)acetate 95%

Dodavatel: Apollo Scientific

Synonyms: (Pyridin-3-yloxy)acetic acid ethyl ester

OR906420-10G OR906420-1G
APOSOR906420-10GEA 6220 CZK
APOSOR906420-10G APOSOR906420-1G
Ethyl 2-(pyridin-3-yloxy)acetate 95%
Ethyl-2-(pyridin-3-yloxy)acetate
Vzorec: C₉H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD00093962
CAS číslo: 18342-98-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR