Tisk…

4-Bromo-7H-pyrrolo[2,3-d]pyrimidine 98%

Dodavatel: Apollo Scientific

OR906413-5G
APOSOR906413-5GEA 4300 CZK
APOSOR906413-5G
4-Bromo-7H-pyrrolo[2,3-d]pyrimidine 98%
4-Bromo-7H-pyrrolo[2,3-d]pyrimidine
Vzorec: C₆H₄BrN₃
MW: 198,0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08445698
CAS číslo: 889939-42-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR