Tisk…

4-Bromo-2-methylpyrimidine 97%

Dodavatel: Apollo Scientific

OR906386-1G OR906386-250MG
APOSOR906386-1GEA 14680 CZK
APOSOR906386-1G APOSOR906386-250MG
4-Bromo-2-methylpyrimidine 97%
4-Bromo-2-methylpyrimidine
Vzorec: C₅H₅BrN₂
Storage Temperature: Ambient
MDL Number: MFCD11223245
CAS číslo: 1114560-76-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR