Tisk…

Diisopropyl phosphonate 95%

Dodavatel: Apollo Scientific

OR906349-100G OR906349-25G
APOSOR906349-100GEA 2700 CZK
APOSOR906349-100G APOSOR906349-25G
Diisopropyl phosphonate 95%
Diisopropyl phosphonate
Vzorec: C₆H₁₅O₃P
MW: 166,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00117905
CAS číslo: 1809-20-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR