Tisk…

2,6-Dichloroimidazo[1,2-b]pyridazine 95%

Dodavatel: Apollo Scientific

OR906339-1G OR906339-250MG
APOSOR906339-1GEA 16960 CZK
APOSOR906339-1G APOSOR906339-250MG
2,6-Dichloroimidazo[1,2-b]pyridazine 95%
2,6-Dichloroimidazo[1,2-b]pyridazine
Vzorec: C₆H₃Cl₂N₃
Storage Temperature: Ambient
CAS číslo: 112581-77-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR