Tisk…
6-Chloro-2,3-dihydro-1H-indole hydrochloride 95%
Dodavatel: Apollo Scientific
OR906327-1G OR906327-250MG OR906327-5G
APOSOR906327-1GEA 9230 CZK
APOSOR906327-1G APOSOR906327-250MG APOSOR906327-5G
6-Chloro-2,3-dihydro-1H-indole hydrochloride 95%
6-Chloro-2,3-dihydro-1H-indole hydrochloride
Vzorec: C₈H₉Cl₂N
MW: 190,0700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09026834
CAS číslo: 89978-84-7