Tisk…

Methyl-5-chloronicotinate 98%

Dodavatel: Apollo Scientific

Synonyms: 5-Chloropyridine-3-carboxylic acid methyl ester, Methyl 5-chloronicotinate

OR906050-10G OR906050-1G OR906050-5G
APOSOR906050-10GEA 12800 CZK
APOSOR906050-10G APOSOR906050-1G APOSOR906050-5G
Methyl-5-chloronicotinate 98%
Methyl-5-chloronicotinate
Vzorec: C₇H₆ClNO₂
MW: 171,5810 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09702467
CAS číslo: 51269-81-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR