Tisk…

4-Methyl-5-nitro-1H-indazole 95%

Dodavatel: Apollo Scientific

OR906049-1G OR906049-250MG
APOSOR906049-1GEA 3410 CZK
APOSOR906049-1G APOSOR906049-250MG
4-Methyl-5-nitro-1H-indazole 95%
4-Methyl-5-nitro-1H-indazole
Vzorec: C₈H₇N₃O₂
MW: 177,1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03787902
CAS číslo: 101420-67-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR