Tisk…

6-Chloro-2-methylquinazolin-4(3H)-one 96%

Dodavatel: Apollo Scientific

OR906046-1G OR906046-5G
APOSOR906046-1GEA 953 CZK
APOSOR906046-1G APOSOR906046-5G
6-Chloro-2-methylquinazolin-4(3H)-one 96%
6-Chloro-2-methylquinazolin-4(3H)-one
Vzorec: C₉H₇ClN₂O
MW: 194,6200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00195330
CAS číslo: 7142-09-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR