1-(Pyrimidin-2-yl)piperidin-4-ol 98%
Dodavatel: Apollo Scientific
OR906032-10G
OR906032-1G
OR906032-5G
APOSOR906032-10GEA
7110
CZK
APOSOR906032-10G
APOSOR906032-1G
APOSOR906032-5G
1-(Pyrimidin-2-yl)piperidin-4-ol 98%
1-(Pyrimidin-2-yl)piperidin-4-ol
Vzorec:
C₉H₁₃N₃O MW: 179,2200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05864742 CAS číslo: 893755-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...