2,5-Dichloro-4-methylpyrimidine 96%
Dodavatel: Apollo Scientific
OR906028-1G
APOSOR906028-1GEA
1990
CZK
APOSOR906028-1G
2,5-Dichloro-4-methylpyrimidine 96%
2,5-Dichloro-4-methylpyrimidine
Vzorec:
C₅H₄Cl₂N₂ MW: 163,0100 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13193656 CAS číslo: 1192064-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...