Tisk…

(4-Chlorophenyl)acetic acid ethyl ester 97%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 4-chlorophenylacetate, Ethyl (4-chlorophenyl)acetate

OR906001-25G OR906001-500G
APOSOR906001-25GEA 0 CZK
APOSOR906001-25G APOSOR906001-500G
(4-Chlorophenyl)acetic acid ethyl ester 97%
(4-Chlorophenyl)acetic acid ethyl ester
Vzorec: (C₆H₄Cl)CH₂COOCH₂CH₃
MW: 198,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00018791
CAS číslo: 14062-24-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR