Tisk…

2-Chloro-3-cyanobenzaldehyde 98%

Dodavatel: Apollo Scientific

OR905987-1G OR905987-5G
APOSOR905987-1GEA 1660 CZK
APOSOR905987-1G APOSOR905987-5G
2-Chloro-3-cyanobenzaldehyde 98%
2-Chloro-3-cyanobenzaldehyde
Vzorec: C₈H₄ClNO
Storage Temperature: Ambient
MDL Number: MFCD08460147
CAS číslo: 165187-24-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR