Tisk…

5-(tert-Butyl)isoxazole-3-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR905958-1G OR905958-5G
APOSOR905958-1GEA 2730 CZK
APOSOR905958-1G APOSOR905958-5G
5-(tert-Butyl)isoxazole-3-carboxylic acid 95%
5-(tert-Butyl)isoxazole-3-carboxylic acid
Vzorec: C₈H₁₁NO₃
MW: 169,1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07377301
CAS číslo: 90607-21-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR