Tisk…

6-Methoxy-3-methyl-1H-indazole 98%

Dodavatel: Apollo Scientific

OR905921-1G OR905921-250MG
APOSOR905921-1GEA 10350 CZK
APOSOR905921-1G APOSOR905921-250MG
6-Methoxy-3-methyl-1H-indazole 98%
6-Methoxy-3-methyl-1H-indazole
Vzorec: C₉H₁₀N₂O
MW: 162,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07780793
CAS číslo: 7746-29-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR