Tisk…

4-Bromobutyryl chloride 97%

Dodavatel: Apollo Scientific

OR905872-100G OR905872-25G
APOSOR905872-100GEA 4300 CZK
APOSOR905872-100G APOSOR905872-25G
4-Bromobutyryl chloride 97%
4-Bromobutyryl chloride
Vzorec: C₄H₆BrClO
Bod varu: 101 °C/37mmHg
Hustota: 1,602 g/cm³ (25 °C)
Bod vzplanutí: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00000753
CAS číslo: 927-58-2
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR