4-tert-Butyl-2-nitrophenol 98%
Dodavatel: Apollo Scientific
Synonyms:
4-(tert-Butyl)-2-nitrophenol
OR905846-100G
OR905846-25G
APOSOR905846-100GEA
5890
CZK
APOSOR905846-100G
APOSOR905846-25G
4-tert-Butyl-2-nitrophenol 98%
4-tert-Butyl-2-nitrophenol
Vzorec:
C₁₀H₁₃NO₃ MW: 195,2151 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00191888 CAS číslo: 3279-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...