7-Chloro-4-chromanone 95%
Dodavatel: Apollo Scientific
OR905815-1G
OR905815-5G
APOSOR905815-1GEA
1870
CZK
APOSOR905815-1G
APOSOR905815-5G
7-Chloro-4-chromanone 95%
7-Chloro-4-chromanone
Vzorec:
C₉H₇ClO₂ MW: 182.6000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09744054 CAS číslo: 18385-72-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...