Tisk…

tert-Butyl-2-methyl-4-oxopiperidine-1-carboxylate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Methylpiperidin-4-one, N-BOC protected, tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate

OR9058-25G
APOSOR9058-25GEA 3910 CZK
APOSOR9058-25G
tert-Butyl-2-methyl-4-oxopiperidine-1-carboxylate 97%
tert-Butyl-2-methyl-4-oxopiperidine-1-carboxylate
Vzorec: C₁₁H₁₉NO₃
MW: 213,2735 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04035608
CAS číslo: 190906-92-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR