3-Amino-1-cyclopentyl-1H-pyrazole-4-carbonitrile 95%
Dodavatel: Apollo Scientific
OR905788-1G
OR905788-250MG
APOSOR905788-1GEA
5060
CZK
APOSOR905788-1G
APOSOR905788-250MG
3-Amino-1-cyclopentyl-1H-pyrazole-4-carbonitrile 95%
3-Amino-1-cyclopentyl-1H-pyrazole-4-carbonitrile
Vzorec:
C9H12N4 MW: 176,2200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11040167 CAS číslo: 122799-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...