(S)-Butyl-2-aminopropanoate 95%
Dodavatel: Apollo Scientific
Synonyms:
H-Ala-OBu, (S)-Butyl 2-aminopropanoate
OR905744-1G
OR905744-5G
APOSOR905744-1GEA
1990
CZK
APOSOR905744-1G
APOSOR905744-5G
(S)-Butyl-2-aminopropanoate 95%
(S)-Butyl-2-aminopropanoate
Vzorec:
C₇H₁₅NO₂ MW: 145,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12796122 CAS číslo: 2885-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...