Tisk…

4-Chloro-6-methyl-2-(methylthio)pyrimidine 95%

Dodavatel: Apollo Scientific

Synonyms: 4-chloro-6-methyl-2-(methylsulphanyl)pyrimidine, 4-Chloro-6-methyl-2-methylthiopyrimidine

OR905698-25G
APOSOR905698-25GEA 3580 CZK
APOSOR905698-25G
4-Chloro-6-methyl-2-(methylthio)pyrimidine 95%
4-Chloro-6-methyl-2-(methylthio)pyrimidine
Vzorec: C₆H₇ClN₂S
MW: 174,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023233
CAS číslo: 17119-73-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR