Tisk…

5,7-Dihydroxyimidazo[1,2-a]pyrimidine 95%

Dodavatel: Apollo Scientific

OR905656-1G OR905656-250MG
APOSOR905656-1GEA 3610 CZK
APOSOR905656-1G APOSOR905656-250MG
5,7-Dihydroxyimidazo[1,2-a]pyrimidine 95%
5,7-Dihydroxyimidazo[1,2-a]pyrimidine
Vzorec: C₆H₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09834820
CAS číslo: 51647-90-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR