Tisk…

5-Bromobenzo[d]isoxazol-3-ylamine 98%

Dodavatel: Apollo Scientific
Varování

OR905603-25G OR905603-5G
APOSOR905603-25GEA 6680 CZK
APOSOR905603-25G APOSOR905603-5G
5-Bromobenzo[d]isoxazol-3-ylamine 98%
5-Bromobenzo[d]isoxazol-3-ylamine
Vzorec: C₇H₅BrN₂O
MW: 213,0300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09834823
CAS číslo: 455280-00-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR