Tisk…

3-Bromo-N,N-dimethylpropan-1-amine hydrobromide 95%

Dodavatel: Apollo Scientific

OR905595-5G
APOSOR905595-5GEA 1700 CZK
APOSOR905595-5G
3-Bromo-N,N-dimethylpropan-1-amine hydrobromide 95%
3-Bromo-N,N-dimethylpropan-1-amine hydrobromide
Vzorec: C5H13Br2N
MW: 246,9714 g/mol
Bod tání: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD11109991
CAS číslo: 5845-30-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR