(4-Chloropyridin-2-yl)methanamine 95%
Dodavatel: Apollo Scientific
OR905574-1G
OR905574-5G
APOSOR905574-1GEA
2200
CZK
APOSOR905574-1G
APOSOR905574-5G
(4-Chloropyridin-2-yl)methanamine 95%
(4-Chloropyridin-2-yl)methanamine
Vzorec:
C₆H₇ClN₂ MW: 142.5900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07374904 CAS číslo: 180748-30-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...