4-Hydroxyphenylpyruvic acid 95%
Dodavatel: Apollo Scientific
Synonyms:
3-(4-Hydroxyphenyl)-2-oxopropanoic acid
OR905458-1G
OR905458-5G
APOSOR905458-1GEA
2820
CZK
APOSOR905458-1G
APOSOR905458-5G
4-Hydroxyphenylpyruvic acid 95%
4-Hydroxyphenylpyruvic acid
Vzorec:
C₉H₈O₄ MW: 180,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002591 CAS číslo: 156-39-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...