Tisk…

Benzyl isocyanide 95%

Dodavatel: Apollo Scientific

OR905439-1G OR905439-5G
APOSOR905439-1GEA 0 CZK
APOSOR905439-1G APOSOR905439-5G
Benzyl isocyanide 95%
Benzyl isocyanide
Vzorec: C₈H₇N
Storage Temperature: Ambient
MDL Number: MFCD00000004
CAS číslo: 10340-91-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR