Tisk…

1-(2-Chlorophenyl)-1H-pyrrole 95%

Dodavatel: Apollo Scientific

OR905437-1G OR905437-250MG
APOSOR905437-1GEA 11180 CZK
APOSOR905437-1G APOSOR905437-250MG
1-(2-Chlorophenyl)-1H-pyrrole 95%
1-(2-Chlorophenyl)-1H-pyrrole
Vzorec: C₁₀H₈ClN
Storage Temperature: Ambient
MDL Number: MFCD02665228
CAS číslo: 89096-75-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR