Tisk…

Methyl-6-methylpicolinate 98%

Dodavatel: Apollo Scientific

OR905393-100G OR905393-25G
APOSOR905393-100GEA 6050 CZK
APOSOR905393-100G APOSOR905393-25G
Methyl-6-methylpicolinate 98%
Methyl-6-methylpicolinate
Vzorec: C₈H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD06203670
CAS číslo: 13602-11-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR