2-Methoxypyrimidine-4-carbaldehyde 97%
Dodavatel: Apollo Scientific
OR905384-1G
OR905384-250MG
APOSOR905384-1GEA
11870
CZK
APOSOR905384-1G
APOSOR905384-250MG
2-Methoxypyrimidine-4-carbaldehyde 97%
2-Methoxypyrimidine-4-carbaldehyde
Vzorec:
C₆H₆N₂O₂ Storage Temperature: Ambient |
CAS číslo:
164738-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...