Tisk…

Benzo[b]thiophene-2-carbonitrile 95%

Dodavatel: Apollo Scientific

OR905297-1G OR905297-5G
APOSOR905297-1GEA 1820 CZK
APOSOR905297-1G APOSOR905297-5G
Benzo[b]thiophene-2-carbonitrile 95%
Benzo[b]thiophene-2-carbonitrile
Vzorec: C₉H₅NS
MW: 159,2077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09751294
CAS číslo: 55219-11-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR