(1-Methyl-1H-benzo[d]imidazol-6-yl)methanol 98%
Dodavatel: Apollo Scientific
OR905132-1G
OR905132-250MG
APOSOR905132-1GEA
17990
CZK
APOSOR905132-1G
APOSOR905132-250MG
(1-Methyl-1H-benzo[d]imidazol-6-yl)methanol 98%
(1-Methyl-1H-benzo[d]imidazol-6-yl)methanol
Vzorec:
C₉H₁₀N₂O Storage Temperature: Ambient |
MDL Number:
MFCD11977685 CAS číslo: 181867-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...