Tisk…

2-Amino-3-(dimethylamino)pyrazine 95%

Dodavatel: Apollo Scientific

OR905054-1G OR905054-250MG
APOSOR905054-1GEA 13230 CZK
APOSOR905054-1G APOSOR905054-250MG
2-Amino-3-(dimethylamino)pyrazine 95%
2-Amino-3-(dimethylamino)pyrazine
Vzorec: C₆H₁₀N₄
Storage Temperature: Ambient
CAS číslo: 89488-74-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR