Tisk…

5-Bromo-3-ethynylpyridin-2-ylamine 95%

Dodavatel: Apollo Scientific

OR904985-1G OR904985-250MG
APOSOR904985-1GEA 5030 CZK
APOSOR904985-1G APOSOR904985-250MG
5-Bromo-3-ethynylpyridin-2-ylamine 95%
5-Bromo-3-ethynylpyridin-2-ylamine
Vzorec: C₇H₅BrN₂
MW: 197,0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566045
CAS číslo: 1210838-82-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR