Tisk…

6-Chloro-5-methylpyridine-2-carboxylic acid 98%

Dodavatel: Apollo Scientific

OR904930-1G OR904930-250MG
APOSOR904930-1GEA 5530 CZK
APOSOR904930-1G APOSOR904930-250MG
6-Chloro-5-methylpyridine-2-carboxylic acid 98%
6-Chloro-5-methylpyridine-2-carboxylic acid
Vzorec: C₇H₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD13176716
CAS číslo: 1166828-13-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR