3-(tert-Butyl)-1H-pyrazole-4-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR904797-1G
OR904797-250MG
APOSOR904797-1GEA
3740
CZK
APOSOR904797-1G
APOSOR904797-250MG
3-(tert-Butyl)-1H-pyrazole-4-carbaldehyde 95%
3-(tert-Butyl)-1H-pyrazole-4-carbaldehyde
Vzorec:
C8H12N2O MW: 152,1937 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07186448 CAS číslo: 1001020-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...